Media for primary cells, melanocyte culture medium M2
Lieferant: PromoCell
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
C-24300EA 375 EUR InStock
C-24300 C-24305 C-24405 C-24400
Media for primary cells, melanocyte culture medium M2
Zellkulturmedien
Melanocyte Culture Medium M2 is a serum-free and PMA (Phorbol Myristate Acetate) free medium optimised for the cultivation of human melanocytes.
Although optimised for primary human cells, it can also be used for equine, porcine, murine, and rat melanocytes. It is also suitable for human pigment epithelial cells, neuronal cells and melanoma cells.
Lieferumfang: The medium is available as medium (ready-to-use), which consists of 1× 500 ml bottle Basal Medium and 1× vial of SupplementMix. Adding the SupplementMix to the Basal Medium results in the complete growth medium. Basal Medium (with or without phenol red) and the SupplementMix can also be purchased separately.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge