Lateral flow assays for the detection of allergens
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
BL603-10 BL608-10 BL607-25 BL604-10 BL607-10 RBL607-10 BL606-10 BL606-25
RBIOBL603-10EA 229 EUR InStock
RBIOBL603-10 RBIOBL608-10 RBIOBL607-25 RBIOBL604-10 RBIOBL607-10 RBIORBL607-10 RBIOBL606-10 RBIOBL606-25
Lateral flow assays for the detection of allergens
Mikrobiologische Testsysteme Antigen-Antikörper-Tests Ag-AK, Lateral Flow
The Lateral Flow Assay is a qualitative detection assay of residues in surface samples obtained from, for eample, production lines. The assay is also applicable for food samples. The assay is applied for the detection of small amounts of contaminations. Contaminations in food possibly elicit hypersensitivity in sensitive persons. Food production lines have to be free of residues to prevent these allergic reactions. The effectiveness of cleaning can be verified by using the lateral flow swabbing method.
Lieferumfang: One determination per strip (25× test strips/kit)
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge