Anti-IL-33 Mouse Monoclonal Antibody [clone: IL33026B]
Lieferant: Cayman Chemical
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
10811-50 10811-100
CAYM10811-50EA 429 EUR InStock
CAYM10811-50 CAYM10811-100
Anti-IL-33 Mouse Monoclonal Antibody [clone: IL33026B]
Antikörper
Type: Primary
Antigen: IL-33
Clonality: Monoclonal
Clone: IL33026B
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge