Arterioclerosis models
Lieferant: Binhold, Paul Lehrmittel
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
1000278 1005611
BINH1000278EA 157 EUR InStock
BINH1000278 BINH1005611
Arterioclerosis models
Modelle Humanbiologie
A horizontally dissected artery fork is depicted with arteriosclerotic changes in four different stages from slightly sedimented to a completely clogged blood vessel.
With this artery model, doctors can explain changes in the blood vessels due to arteriosclerosis.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge