Capillary columns, Stabilwax®-DA (fused silica)
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
11005 11007 11009 11020 11021 11022 11023 11024 11025 11026 11027 11028 11035 11036 11037 11038 11039 11040 11041 11042 11043 11051 11052 11055 11057 11058 11062 11065 11068
REST11005EA 486 EUR InStock
REST11005 REST11007 REST11009 REST11020 REST11021 REST11022 REST11023 REST11024 REST11025 REST11026 REST11027 REST11028 REST11035 REST11036 REST11037 REST11038 REST11039 REST11040 REST11041 REST11042 REST11043 REST11051 REST11052 REST11055 REST11057 REST11058 REST11062 REST11065 REST11068
Capillary columns, Stabilwax®-DA (fused silica)
Chromatographiesäulen
Application specific columns for free (underivatized) acids and some inorganic acids. Resistant to oxidative damage.
Polar phase; Crossbond® acid deactivated Carbowax® polyethylene glycol for acidic compounds
Stabilwax®-DA bonded polyethylene glycol has an acidic functionality incorporated into the polymer structure. This permits analysis of acidic compounds without derivatisation, significantly reduces adsorption of acids and increases sample capacity for volatile free acids. These columns give better peak shapes for high molecular weight acids.
Temperature range: 40 to 260 °C
Similar phases: HP-FFAP, DB-FFAP, CP-WAX 58 FFAP CB, NUKOL, ZB-FFAP. Equivalent to USP G25, G35 phases.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge