Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-V5 Rabbit Polyclonal Antibody (Biotin)

Lieferant: Immunoreagents

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

Sortieren nach:

IMMRIR2857EA 469 EUR InStock
IMMRIR2857
Anti-V5 Rabbit Polyclonal Antibody (Biotin)
The ImmunoReagents' NHS-LC-biotin (succinimidyl-6-(biotinamindo)hexanoate), also known as NHS-X-biotin is a derivative of D-biotin, a amine-reactive biotinylation agent, that contains a spacer arm off the valeric acid side chain of D-biotin with an NHS ester group at its end. The NHS ester group at the end of NHS-LC-biotin covalently binds to amine groups in proteins and other molecules forming a stable amide linkage and releasing the NHS group. The 6-aminocaproic acid spacer of NHS-LC-biotin greatly increases the length between a covalently modified molecule and the bicyclic biotin rings leading to a better binding potential for avidin or streptavidin probes. NHS-LC-biotin is insoluble in aqueous environments requiring the dissolution of organic solvents prior to the addition to a buffered reaction.

Type: Tag
Antigen: V5
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge