Capillary adaptable FID jet
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
20672 20670 20671 20673
REST20672EA 118 EUR InStock
REST20672 REST20670 REST20671 REST20673
Capillary adaptable FID jet
Chromatographie-Zubehör Chromatographie-Ersatzteile
These capillary adaptable FID replacement jets are engineered with a fluted tip to guide the capillary column into the jet.
  • For Agilent 5890/6850/6890 GCs
  • Available in standard or Siltek treated high-performance versions
Threads in standard version are specially coated for easy installation and removal, also special processing ensures the highest degree of cleanliness.
Int.Ø×L: 0,28×61,5 mm.
The high-performance versions are siltek treated which are extremely inert for use with active compounds.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge