Back pressure regulator
Lieferant: Perkin Elmer
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
PERC02540272EA 0 EUR InStock
PERC02540272
Back pressure regulator
Ventile Rückschlagventile
This back pressure regulator can be used to generate backpressure for applications less than 250 psi.
Order Now
PRODUKT
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge