40503.22
40503.36
40503.22EA
43.2
EUR
InStock
40503.22
40503.36
Magnesium chromate hydrate ≥99,8% (Metall-Basis)
Magnesium chromate hydrate
Purity: 99.8%
CAS No.: 16569-85-0
Molecular Formula: MgCrO4.xH2O
Formula Weight: 140.30(anhy)
Physical Form: Powder
Solubility: Soluble in water
UN No.: UN2923
MDL No.: MFCD00801009
Applications: Corrosion inhibitor, pigment