88322.22
88322.36
88322.22EA
109
EUR
InStock
88322.22
88322.36
Aluminum silicon ≥99% (Metall-Basis), Pulver -325 mesh
Aluminum silicon
Purity: 99%
CAS No.: 11145-27-0
Molecular Formula: Al:Si; 88:12 wt%
Physical Form: Powder
Odour: Odourless
UN No.: UN1398
MDL No.: MFCD00143595