H27238.03
H27238.06
H27238.03EA
53
EUR
InStock
H27238.03
H27238.06
Tris(cyclopentadienyl)terbium(III) 99% (99,9% Tb)
Tris(cyclopentadienyl)terbium(III)
Purity: 99%
CAS No.: 1272-25-9
Molecular Formula: C15H15Tb
Formula Weight: 354.22
Melting Point: 235°
UN No.: UN3181
MDL No.: MFCD03427139