L10902.03
L10902.06
L10902.03EA
44.4
EUR
InStock
L10902.03
L10902.06
4-Vinylphenol -Lösung ≥10% in Propylenglykol
4-Vinylphenol
CAS No.: 2628-17-3
Molecular Formula: C8H8O
Formula Weight: 120.15
Storage Temperature: -30°C to -10°C
Flash Point: 100°(212°F)
Refractive Index: 1.4470
Beilstein Reference: 506844
MDL No.: MFCD00017593