L11886.MC
L11886.ME
L11886.MCEA
125
EUR
InStock
L11886.MC
L11886.ME
Myosmine, (max. 2% H₂O) 98%
Myosmine
Purity: 98%
CAS No.: 532-12-7
Molecular Formula: C9H10N2
Formula Weight: 146.19
Melting Point: 42-44°
Boiling Point: 82-83°/0.5mm
Beilstein Reference: 119254
MDL No.: MFCD00052019