L16848.22
L16848.06
L16848.14
L16848.22EA
240
EUR
InStock
L16848.22
L16848.06
L16848.14
Perfluorotriamylamine ≥85%, tech.
Perfluorotriamylamine
Purity: 85%
CAS No.: 338-84-1
Molecular Formula: [CF3(CF2)4]3N
Formula Weight: 821.12
Boiling Point: 210-220°
MDL No.: MFCD00042367