Streptavidin agarose, Pierce™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
20349 20347 20351 20353
PIER20349EA 674 EUR InStock
PIER20349 PIER20347 PIER20351 PIER20353
Streptavidin agarose, Pierce™
Protein-Aufbereitungs Protein Purification Resins
Pierce™ Streptavidin agarose is a standard-capacity crosslinked 6% beaded agarose resin of immobilised recombinant streptavidin protein for use in a variety of biotin-binding affinity purification methods.
  • Purified recombinant streptavidin protein (53 kDa, near-neutral pI); tetrameric with four biotin-binding site per molecule
  • Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Streptavidin is immobilised by charge-free, leach-resistant covalent bonds, resulting in low non specific binding and enabling multiple uses without decline in yield
  • Standard capacity beads with a normal load of immobilised streptavidin, providing a binding capacity of 1 to 3 mg of biotinylated BSA per ml of resin
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge