Fluor phosphine reagents, DyLight™, Pierce™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
88907 88910 88911
PIER88907EA 582 EUR InStock
PIER88907 PIER88910 PIER88911
Fluor phosphine reagents, DyLight™, Pierce™
Proteinkonjugate und Markierungen
DyLight™ Fluor phosphine reagents are phosphine-activated fluorescent dyes for specific labeling and detection of azide-tagged molecules, which enables use of fluorescence imaging in metabolic labeling strategies.
  • Easily dissolves in water-miscible solvents (eg. DMSO) for subsequent dilution in aqueous reaction mixtures with cell lysates and other biological samples
  • Reaction chemistry occurs effectively in simple buffer conditions requiring no accessory reagents such as copper or reducing agents, and does not interfere with fluorescence applications
  • The phosphine reactive group is specific in biological samples for bioorthogonal azide-tagged molecules, ensuring that fluorescent labeling is specific
  • DyLight™ 488, 550 and 650 are intense, highly stable fluorophores for green, orange and red fluorescent detection
When used in combination with azide labeling strategies, phosphine-activated dyes enable selective fluorescent labeling for detection of protein interactions and post-translational modifications using fluorescence imaging technologies. The phosphine group conjugates to azide groups by the Staudinger reaction mechanism.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge
Diese Webseite verwendet Cookies, eigene oder von Drittanbieter, zur Sammlung statistischer Informationen, um Inhalte und Anzeigen für Sie zu personalisieren. Wenn Sie diese Seite weiterhin verwenden, akzeptieren Sie die Cookie-Nutzung.
Weitere Informationen finden Sie in unseren Datenschutzbestimmungen..