Immobilised Protein G, UltraLink™, Pierce™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
53125 53128 53126 53127
PIER53125EA 468 EUR InStock
PIER53125 PIER53128 PIER53126 PIER53127
Immobilised Protein G, UltraLink™, Pierce™
Protein-Aufbereitungs Protein Purification Resins
Pierce™ Protein G UltraLink™ and Protein G Plus UltraLink™ resins are useful for purification of antibodies and immunoprecipitation (IP). Samples containing IgG are incubated with the resin-immobilised Protein G in a buffer that facilitates binding. After non-IgG and non-antigen components of the sample are washed from the resin, the bound IgG and antigen may be recovered by elution.
  • Immobilised Protein G specifically binds IgG from several mammalian species, especially all isotypes of IgG from human, mouse and rat
  • Acrylamide based support provides an alternative to crosslinked beaded agarose supports for medium pressure applications
Protein G UltraLink™ resin binding capacity: >20 mg human IgG/ml resin.
Protein G UltraLink™ column binding capacity: >40 mg human IgG/ml resin.
Protein G Plus UltraLink™ resin binding capacity: >25 mg human IgG/ml resin.
Lieferumfang: Available as resin or as Protein G UltraLink Columns (2×2 ml resin pre-packed in columns with twist-off bottom tabs and an accessory pack containing two white tips).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge
Diese Webseite verwendet Cookies, eigene oder von Drittanbieter, zur Sammlung statistischer Informationen, um Inhalte und Anzeigen für Sie zu personalisieren. Wenn Sie diese Seite weiterhin verwenden, akzeptieren Sie die Cookie-Nutzung.
Weitere Informationen finden Sie in unseren Datenschutzbestimmungen..