PLOT columns, Rtx®-U-bond columns (fused silica)
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
19752 19750
REST19752EA 865 EUR InStock
REST19752 REST19750
PLOT columns, Rtx®-U-bond columns (fused silica)
Chromatographiesäulen
Highly polar, divinyl benzene ethylene glycol/dimethylacrylate.
  • Ideal for analysis of polar and nonpolar compounds
  • Ideal for trace H₂S, COS, and mercaptans in hydrocarbon streams
Temperature range: Stable to 190 °C.
Similar phases: HP-PLOT U, CP-PoraPLOT U and CP-PoraBOND U.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge