Schläuche, Silikon, Sterilin™
Lieferant: Sterilin
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
TSR0600200P TSR0400200P TSR0500160P TSR0320100P
SLINTSR0600200PEA 187 EUR InStock
SLINTSR0600200P SLINTSR0400200P SLINTSR0500160P SLINTSR0320100P
Schläuche, Silikon, Sterilin™
Schläuche Transferschlauch
Sterilin™ silicone tubing is non-toxic and ideal for medical, pharmaceutical and food applications with high quality, nontoxic Thermo Scientific™ Sterilin™ Silicone Tubing. Not available in all geographic regions.
  • Laser etched batch code for ease of traceability
  • Ideal for use with peristaltic pumps
  • Can be autoclaved or dry heat sterilised
  • 15 m length
  • Aseptically manufactured
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge