Disposable tip adapters
Lieferant: HAMILTON BONADUZ
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
31330 31331
HAMI31330EA 41.1 EUR InStock
HAMI31330 HAMI31331
Disposable tip adapters
Spritzen Spritzen für die Chromatographie
These adapters enable the connection of a standard 1 ml or 200 µl disposable pipette tip to the end of a Hamilton syringe. The adapter connects to standard Luer Tip (LT) and PTFE Luer Lock (TLL) syringes. When used in conjunction the PB600-1 and the disposable tip adapters can be used as a small volume repeating pipette.
  • Ideal for eliminating the possibility for cross contamination
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge