Certified calibration standard kits for spectrophotometer, OQ/PV
Lieferant: AGILENT HEWLETT PACKARD
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
5063-6503 5063-6521
HEWL5063-6503EA 0 EUR InStock
HEWL5063-6503 HEWL5063-6521
Certified calibration standard kits for spectrophotometer, OQ/PV
Standards Farbreferenzen und Spektroskopie-Standards
The chemical kits can be used with any UV-Vis system and consist of pre-prepraed solutions in sealed ampoules.
  • Each standard includes a Certificate of Analysis for traceability
  • Solutions are specified by United States Pharmacopeia (USP) and European Pharmacopeia (EP)
  • Designed to conform both quality and regulatory requirements when performing UV-visible measurements
Solutions include holmium oxide for wavelength accuracy measurement. Potassium dichromate for photometric accuracy measurement. Sodium nitrite, sodium iodide and potassium chloride for stray light measurements at 340, 220 and 200 nm; and toluene in hexane for resolution measurement.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge