BrightVision Anti-IgG (H+L) (Ultimate kit)
Lieferant: IMMUNOLOGIC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
UBVO110HRP UBVO500HRP UBVO999HRP
UBVO110HRPEA 1160 EUR InStock
UBVO110HRP UBVO500HRP UBVO999HRP
BrightVision Anti-IgG (H+L) (Ultimate kit)
Antikörper
BrightVision immunohistostaining reagents utilise a novel controlled polymerisation technology to prepare polymeric HRP linker antibody conjugates. Comparing to conventional biotin-streptavidin based detection kits, these kits have the advantages of higher amplification power, biotin free and more consistent immunostaining outcomes on archival tissues and on difficult-to-work antibodies. These advantages would bring to laboratories the benefit of more accurate result, faster turn-around, less trouble shooting and better costs-saving. The polymer complex will be visualized with the Bright DAB appropriate substrate or chromogen.
  • High-quality secondary antibody and substrate for IHC applications
  • Antigen specificity: Anti-Mouse IgG (H+L), Anti-Rabbit IgG (H+L)
  • Kit contains poly-HRP-anti-mouse/rabbit IgG (ready-to-use) and Bright DAB (substrate + chromogen)
Zertifizierungen: Meets CE/IVD standards.
Lieferumfang: Shipped with blue ice. Store between 2 to 8 °C.
Achtung! Bitte beachten: DAB is a suspected carcinogen. Do not pipet by mouth. Avoid contact with skin and eyes. DAB can be deactivated with equal parts of Clorox.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge