Transfer plasmids, pOET Insect and BacMam
Lieferant: MIRUS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
MIR6150 MIR6151 MIR6152
MIRUMIR6150EA 484 EUR InStock
MIRUMIR6150 MIRUMIR6151 MIRUMIR6152
Transfer plasmids, pOET Insect and BacMam
Transfektionsreagenzien
The pOET Insect and BacMam transfer plasmids for high yield protein production in insect and mammalian cells are designed for use with the flashBAC™ baculovirus expression system and TransIT®-Insect transfection reagent.
  • Simple construction of recombinant virus when used with the flashBAC™ baculovirus expression system
  • Transfer plasmids are available for insect or mammalian cell expression
  • Can be used with any baculovirus expression system that relies on recombination in insect cells at the polyhedrin locus
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge