HPLC columns, Brownlee™ for HRes™ fast-LC IBD
Lieferant: Perkin Elmer
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
Brownlee™
PERCN9303921EA 851 EUR InStock
PERCN9303921 PERCN9303920 PERCN9303922
HPLC columns, Brownlee™ for HRes™ fast-LC IBD
Intrinsically base-deactivated (IBD) phase, containing a polar group within, or intrinsic to, the hydrocarbon bonded phase. Unique selectivity and a high level of base deactivation, while reducing or eliminating the need for mobile phase additives.
  • Packed to withstand high operating pressures
Excellent phase for significantly increased retention and selectivity of acids, symmetrical peak shapes of bases and for the analysis of zwitterions and other polar compounds.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge