HPLC conventional columns, Brownlee™ analytical C18
Lieferant: Perkin Elmer
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
Brownlee™
PERCN9303509EA 531 EUR InStock
PERCN9303509 PERCN9303508 PERCN9303677 PERCN9303512 PERCN9303864 PERCN9303511 PERCN9303863 PERCN9303676 PERCN9303866 PERCN9303514 PERCN9303513 PERCN9303865 PERCN9303868 PERCN9303867 PERCN9303507 PERCN9303862 PERCN9303510 PERCN9303861
HPLC conventional columns, Brownlee™ analytical C18
These Brownlee analytical columns are built on 110 Å pore size type A silica in 3 and 5 μm particle sizes that is equivalent to the classic Hypersil® silica. Provide excellent peak shape for acidic and neutral analytes but are generally not recommended for basic analytes.
  • With intermediate carbon loading and surface area
  • Suitable for a wide range of acidic to neutral hydrophobic compounds
These columns provide reliable performance and excellent peak symmetry for a broad range of acidic and neutral hydrophobic compounds.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge