UHPLC columns, Ultra IBD (USP L68)
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
9175312-700 9175313-700 9175315-700 9175332-700 9175333-700 9175335-700 9175352-700 9175353-700 9175355-700 9175362-700 9175363-700 9175365-700 9175512-700 9175513-700 9175515-700 9175522-700 9175523-700 9175525-700 9175532-700 9175533-700 9175535-700 9175552-700 9175553-700 9175555-700 9175562-700 9175563-700 9175565-700 9175572-700 9175573-700 9175575-700 917531E 917533E 917535E 917536E 917551E 917552E 917553E 917555E 917556E 917557E
REST9175312-700EA 699 EUR InStock
REST9175312-700 REST9175313-700 REST9175315-700 REST9175332-700 REST9175333-700 REST9175335-700 REST9175352-700 REST9175353-700 REST9175355-700 REST9175362-700 REST9175363-700 REST9175365-700 REST9175512-700 REST9175513-700 REST9175515-700 REST9175522-700 REST9175523-700 REST9175525-700 REST9175532-700 REST9175533-700 REST9175535-700 REST9175552-700 REST9175553-700 REST9175555-700 REST9175562-700 REST9175563-700 REST9175565-700 REST9175572-700 REST9175573-700 REST9175575-700 REST917531E REST917533E REST917535E REST917536E REST917551E REST917552E REST917553E REST917555E REST917556E REST917557E
UHPLC columns, Ultra IBD (USP L68)
Chromatographiesäulen
These are stationary phase columns made with high-purity and type-B silica that minimises activity and creates high-density bonding.
These columns are widely used for reliable HPLC applications.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge