Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ICAM3 Mouse Monoclonal Antibody (CF647) [clone: 101-1D2]

Lieferant: Biotium

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

Sortieren nach:

BNC470177-500 BNC470177-100
BNC470177-500EA 560 EUR InStock
BNC470177-500 BNC470177-100
Anti-ICAM3 Mouse Monoclonal Antibody (CF647) [clone: 101-1D2]
Antikörper
Recognizes an N-glycosylated glycoprotein of 120 kDa with intra-chain disulfide bonds, identified as CD50 or ICAM-3 (WS: IV & V). Its epitope localizes in the D2 extracellular domain and is resistant to neuraminidase and proteases. CD50 is the major ligand for LFA-1 (CD11a/CD18) and may have signalling role to increase adhesion. It is expressed on thymocytes and T lymphocytes and is resistant to treatment with phosphatidylinositol (PI) phospholipase C. This MAb inhibits primary mixed lymphocyte culture (MLC) but not secondary MLC, cytotoxicity or proliferation induced by mitogens. It blocks binding of NK1-L16 stimulated T cells to L cells expressing CD50. This MAb is excellent for staining of formalin/paraffin tissues.

Type: Primary
Antigen: ICAM3
Clonality: Monoclonal
Clone: 101-1D2
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge