Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TLR2 Mouse Monoclonal Antibody (CF488A) [clone: TLR2/221]

Lieferant: Biotium

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

Sortieren nach:

BNC880221-500 BNC880221-100
BNC880221-500EA 560 EUR InStock
BNC880221-500 BNC880221-100
Anti-TLR2 Mouse Monoclonal Antibody (CF488A) [clone: TLR2/221]
Antikörper
This MAb reacts with human Toll-like receptor 2 (TLR2). It is a member of the Toll-like receptor (TLR) family, which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. This gene is expressed most abundantly in peripheral blood leukocytes, and mediates host response to Gram-positive bacteria and yeast via stimulation of NF-kappaB.

Type: Primary
Antigen: TLR2
Clonality: Monoclonal
Clone: TLR2/221
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Dog, Human, Monkey

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge