Anti-VIM Mouse Monoclonal Antibody (CF488A) [clone: VM452]
Lieferant: Biotium
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
BNC880452-500 BNC880452-100
BNC880452-500EA 560 EUR InStock
BNC880452-500 BNC880452-100
Anti-VIM Mouse Monoclonal Antibody (CF488A) [clone: VM452]
Antikörper
This MAb reacts with a 58 kDa protein identified as vimentin. It shows no cross-reaction with other closely related intermediate filament proteins (IFP's) such as desmin, keratin, neurofilament, and glial fibrillary acid protein.Anti-vimentin alone is of limited value as a diagnostic tool; however, when used in panels with other antibodies, it is useful for the sub-classification of a given tumor. Expression of vimentin, when used in conjunction with anti-keratin, is helpful when distinguishing melanomas from undifferentiated carcinomas and large cell lymphomas. All melanomas and Schwannomas react strongly with anti-vimentin. It labels a variety of mesenchymal cells, including melanocytes, lymphocytes, endothelial cells, and fibroblasts. Non-reactivity of anti-vimentin is often considered more useful than its positive reactivity, since there are a few tumors that do not contain vimentin, e.g. hepatoma and seminoma. Anti-vimentin is also useful as a tissue process control reagent.
Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: VM452
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cat, Chicken, Cow, Dog, Goat, Human, Pig
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge