Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FCGR3A Mouse Monoclonal Antibody (CF488A) [clone: HuNK2]

Lieferant: Biotium

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

Sortieren nach:

BNC880928-500 BNC880928-100
BNC880928-500EA 560 EUR InStock
BNC880928-500 BNC880928-100
Anti-FCGR3A Mouse Monoclonal Antibody (CF488A) [clone: HuNK2]
Antikörper
Recognizes a protein of 50-65 kDa, identified as CD16 (Workshop IV; Code N39 ) (also known low affinity Fc receptor III for IgG (FcRIII) or Leu 11). CD16 exists as a polypepetide-anchored from (FCRIIIA or CD16A) on human natural killer (NK) cells and monocytes/ macrophages and as a glycosylphosphatidylinositol (GPI)-anchored form (FcRIIIB or CD16B) on neutrophils. CD16B is polymorphic and the two alleles are termed NA1 and NA2.3 CD16 plays a role in signal transduction, NK cell activation and antibody-dependent cellular cytotoxicity. This MAb has been showed to inhibit the binding of immune complex to NK cells, inhibit cytotoxicity of NK cells, and induce calcium fluxes in NK cells and neutrophils.

Type: Primary
Antigen: FCGR3A
Clonality: Monoclonal
Clone: HuNK2
Conjugation: CF488A
Epitope:
Host: Mouse
Reactivity: Human
Isotype: IgG2a, kappa

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge