Anti-Speedy/Ringo Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
NB100-2521 NB100-2521SS
NOVUNB100-2521EA 591 EUR InStock
NOVUNB100-2521 NOVUNB100-2521SS
Anti-Speedy/Ringo Rabbit Polyclonal Antibody
Antikörper
Rabbit Polyclonal Speedy/Ringo Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse.
Type: Primary
Antigen: Speedy/Ringo
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge