Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CD3E Mouse Monoclonal Antibody (CF568) [clone: CRIS-7]

Lieferant: Biotium

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

Sortieren nach:

BNC681050-500 BNC681050-100
BNC681050-500EA 560 EUR InStock
BNC681050-500 BNC681050-100
Anti-CD3E Mouse Monoclonal Antibody (CF568) [clone: CRIS-7]
Antikörper
Recognizes the epsilon-chain of CD3 (Workshop V; Code: CD03.09), which consists of five different polypeptide chains (designated as γ, δ, ε, ζ, and η) with MW ranging from 16-28 kDa. The CD3 complex is closely associated at the lymphocyte cell surface with the T cell antigen receptor (TCR). Reportedly, CD3 complex is involved in signal transduction to the T cell interior following antigen recognition. The CD3 antigen is first detectable in early thymocytes and probably represents one of the earliest signs of commitment to the T cell lineage. In cortical thymocytes, CD3 is predominantly intra-cytoplasmic. However, in medullary thymocytes, it appears on the T cell surface. CD3 antigen is a highly specific marker for T cells, and is present in majority of T cell neoplasms.

Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: CRIS-7
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human, Monkey

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge