Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CD38 Mouse Monoclonal Antibody (CF640R) [clone: AT2]

Lieferant: Biotium

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

Sortieren nach:

BNC401086-500 BNC401086-100
BNC401086-500EA 560 EUR InStock
BNC401086-500 BNC401086-100
Anti-CD38 Mouse Monoclonal Antibody (CF640R) [clone: AT2]
Antikörper
This MAb reacts with a 45 kDa glycopeptide, which is a type II membrane glycoprotein with a transmembrane sequence near the NH2terminus. CD38 is a type II transmembrane glycoprotein that is present on early B- and T-cell lineages and activated B- and T-cells but is absent from most mature resting peripheral lymphocytes. CD38 is also found on thymocytes, pre-B cells, germinal center B-cells, mitogen-activated T-cells, monocytes and Ig-secreting plasma cells. CD38 is expressed on CD34 cells. The CD34 CD38- population of hematopoietic stems cells defines the most pluripotent cells (e.g. blast colony forming cells).

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: AT2
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge