GC columns, Rt®-TCEP (fused silica)
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
10998 10999
REST10998EA 777 EUR InStock
REST10998 REST10999
GC columns, Rt®-TCEP (fused silica)
Chromatographiesäulen
General purpose Rt®-TCEP columns is ideal for aromatics and oxygenates in gasoline.
  • Highly polar phase; 1,2,3-tris[2-cyanoethoxy]propane (not bonded)
  • Temperature range: 0 to 150 °C
  • Enhanced polymer stability and extended column lifetime
The extremely polar Rt®-TCEP stationary phase provides a retention index for benzene greater than 1,100 and permits the separation of alcohols and aromatics from the aliphatic constituents in gasoline. These columns can separate a wide variety of compounds with an elution pattern unattainable using other high-polarity siloxanes. Solvent rinsing should be avoided.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge