GC columns, ShinCarbon ST (packed and micropacked)
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
80486-800 80486-810 80486-820 80486-840 19808 19809 19810 19043
REST80486-800EA 906 EUR InStock
REST80486-800 REST80486-810 REST80486-820 REST80486-840 REST19808 REST19809 REST19810 REST19043
GC columns, ShinCarbon ST (packed and micropacked)
Chromatographiesäulen
These ShinCarbon ST columns are made of SilcoSmooth stainless steel material. Analyses oxygen, nitrogen, methane, carbon monoxide and carbon dioxide with one column at room temperature and separate permanent gases, including carbon monoxide and carbon dioxide, without cryogenic cooling.
  • Rapid separations of permanent gas/light hydrocarbon mixtures
  • Compatibile with most GC detectors - minimal bleed, minimal baseline rise
  • Preconditioned, less than 30 minutes to stabilise
  • Maximum temperature of 280/300 °C
ShinCarbon ST material, a high surface area carbon molecular sieve (~1500 m²/g), is the ideal medium for separating gases and highly volatile compounds by gas solid chromatography (GSC). Low-bleed operation and rapid stabilisation. Its 300 °C upper programmed temperature limit minimises bleed and baseline rise during temperature programming, making the material compatible with most detection systems used for gas analysis, including TCD or HID.
Bestellinformation: Columns not include column nuts and ferrules. Optional installation kits can be ordered separately.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge