Replacement chemical traps and parts
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
21610 22820 22800 23031 23032
REST21610EA 210 EUR InStock
REST21610 REST22820 REST22800 REST23031 REST23032
Replacement chemical traps and parts
Vakuumfallen
These replacement chemical traps and parts for Agilent GC are easy to install, attach to same fittings as original manufacturer's equipment.
  • Adsorbents remove both moisture and hydrocarbons
  • Built-in frits retain fine particles
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge