Capillary dedicated FID replacement jet
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
21620 21621 21682 21683 23078
REST21620EA 97 EUR InStock
REST21620 REST21621 REST21682 REST21683 REST23078
Capillary dedicated FID replacement jet
Chromatographie-Zubehör Chromatographie-Ersatzteile
These capillary dedicated FID replacement jet for Agilent 5890/6850/6890/7890 GCs are available in standard, high-temperature or Siltek treated high-performance versions.
  • For standard version threads are specially coated for easy installation and removal
  • Special processing ensures the highest degree of cleanliness
  • High-performance version - Siltek treated are extremely inert for use with active compounds
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge