Direct injection liners, Uniliner®
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
20964 20965 20966
REST20964EA 0 EUR InStock
REST20964 REST20965 REST20966
Direct injection liners, Uniliner®
Chromatographie-Zubehör Chromatographie-Liner
Direct injection Uniliner® liners minimise solvent and peak tailing. These are suitable for 6,35 mm packed column injection ports.
  • Press-Tight® taper forms dead volume free connection to columns
  • Can be used with 0,25, 0,32 and 0,53 mm ID capillary columns
  • Can be removed from the septum nut weldment
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge