Bovine native alpha-crystallin
Lieferant: STRESSGEN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
SPP-225J SPP-225L
STSSSPP-225JEA 0 EUR InStock
STSSSPP-225J STSSSPP-225L
Bovine native alpha-crystallin
Proteine und Peptide
Alpha-crystallins (also known as CRYA, HspB, Heat shock protein β) composed of alpha A (~20 kDa) and alpha B (~20 kDa) subunits, are major water-soluble proteins accounting for almost 50% of total protein in the mammalian transparent eye lens and they are also found in a variety of other tissues.
  • Isolated from bovine eye lens
  • Purified by multi-step chromatography (SDS-PAGE, western blot)
  • Recommended dilutions (conditions) include western blot (100 ng/lane, colorimetric)
It has a liquid formulation in PBS at pH 7,2 containing 0,15M sodium chloride, 0,05M phosphate buffer and 0,09% sodium azide.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge