O-rings, graphite
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
20296 20297 20298 20299
REST20296EA 72 EUR InStock
REST20296 REST20297 REST20298 REST20299
O-rings, graphite
Versiegeln Dichtringe
These O-rings are made of graphite. Ideal for Bruker/Varian 1177 injectors.
  • Maximum operating temperature 450 °C
  • Can used with split or splitless liners
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge