Photoionisation lamps
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
23020 20675 20676
REST23020EA 982 EUR InStock
REST23020 REST20675 REST20676
Photoionisation lamps
Beleuchtung UV-Lampen
Photoionisation (PID) lamps are vacuum tubes designed to emit ultraviolet energy.
Model 108-10.0/10.6: Features both 10,0 and 10,6 eV potential, has a 0,781" base diameter and is used in Tracor, OI and Baseline instruments.
Model 103: Has a 34,9 mm base and is used in HNU and SRI detectors.
Model 108-BTEX: Lamp’s higher output makes it ideal for detection of BTEX compounds.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge