Guard/retention gap columns, polar-deactivated (fused silica)
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
10065 10066 10067 10068 10069 10070 10014 10034
REST10065EA 80 EUR InStock
REST10065 REST10066 REST10067 REST10068 REST10069 REST10070 REST10014 REST10034
Guard/retention gap columns, polar-deactivated (fused silica)
Chromatographiesäulen
Polyethylene glycol deactivation layer provides optimum wettability for polar compounds.
  • Tested with a comprehensive test mix to ensure high inertness
  • Minimize peak splitting when using polar solvents such as methanol or water
  • Compatible with Stabilwax, Rtx-225, and Rt-2330 capillary columns
Maximum temperature: 280 °C
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge