Guard/retention gap columns, intermediate-polar deactivated (passivated stainless steel)
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
70044 70045 70046 70047
REST70044EA 144 EUR InStock
REST70044 REST70045 REST70046 REST70047
Guard/retention gap columns, intermediate-polar deactivated (passivated stainless steel)
Chromatographiesäulen
Useful for a wide range of applications.
  • Compatible with most common solvents
Maximum temperature: 325 °C
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge