Guard/retention gap columns, base-deactivated (fused silica)
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
10000 10001 10002 10000-600 10001-600 10002-600
REST10000EA 88 EUR InStock
REST10000 REST10001 REST10002 REST10000-600 REST10001-600 REST10002-600
Guard/retention gap columns, base-deactivated (fused silica)
Chromatographiesäulen
Recommended for use with Rtx®-5 Amine, Rtx®-35 Amine, Rtx®-Volatile Amine, and Stabilwax®-DB capillary columns.
  • Tested with a basic amine test mix
  • Excellent inertness for basic compounds
  • Batch test chromatogram included
Maximum temperature: 315 °C
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge