Starburst plugs for shell vials
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
21769 21770 21771 21772 21773 21774
REST21769EA 21 EUR InStock
REST21769 REST21770 REST21771 REST21772 REST21773 REST21774
Starburst plugs for shell vials
Verschlüsse Chromatographieampullen-Verschlüsse
These clear Starburst snap plugs are made of PE. Suitable for shell vials, 1 ml WISP 96 style, 8×40 mm.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge