Powder lances
Lieferant: Sampling Systems
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
2250A-1000 2250A-2000 2250A-600
SAMP2250A-1000EA 677 EUR InStock
SAMP2250A-1000 SAMP2250A-2000 SAMP2250A-600
Powder lances
Probenehmer Probenehmer für Feststoffe
The powder lance can be used to manually take large volume samples.
  • Cross sectional sampling without damage
  • Mostly used with cohesive powders
  • Horizontal use through a sack or elsewhere vertically
Suitable for use in the pharmaceutical industries.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge