Internal reducing union, zero dead volume, Valco®
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
20151 20152 20153
REST20151EA 75 EUR InStock
REST20151 REST20152 REST20153
Internal reducing union, zero dead volume, Valco®
Fittings und Verbinder
Internal reducing union used to connect two sizes of tubing, using zero-dead-volume fittings on each end. Suitable for 1,6 to 3,2 mm OD tubing.
Lieferumfang: Supplied with stainless steel ferrules.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge