Internal cross, zero dead volume, Valco®
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
20156 20157
REST20156EA 147 EUR InStock
REST20156 REST20157
Internal cross, zero dead volume, Valco®
Fittings und Verbinder
These internal cross are made of stainless steel. Used to connect four lines. Ideal for 1,6 mm OD tubing.
Lieferumfang: Supplied with stainless steel ferrules.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge