Cross-disc inlet seals for Agilent GC's
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
21009 21010 21011 21012 20474 20475 20476 20477
REST21009EA 87 EUR InStock
REST21009 REST21010 REST21011 REST21012 REST20474 REST20475 REST20476 REST20477
Cross-disc inlet seals for Agilent GC's
Versiegeln Mechanische Dichtungen
Cross disc inlet seals are ideal for high-flow split applications >500 ml/min on Agilent 5890 GCs.
  • Siltek® treatment provides inertness similar to fused silica
The inlet seal at the base of the Agilent 5890/6890/7890 GC injection port contacts the sample and must be changed frequently to prevent adsorption of active compounds. In addition, septum fragments and sample residue accumulate on the disk surface, requiring disk replacement. To reduce breakdown and adsorption of active compounds, Siltek® or gold-plated seals are recommended.
Lieferumfang: All seals include washers.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge