Cables for Dissolved Oxygen and pH Sensors
Lieferant: Mettler - Toledo
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
52300109 52300110 52300108 52300107
662-0613EA 264.25 EUR InStock
662-0613 662-0614 662-0045 METN52300107
Cables for Dissolved Oxygen and pH Sensors
Kabel
These VP patch cables are used for connections between pH/DO sensors and M300 instruments. VP cables are for VP style sensors only.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge